ContactUsLinkComponent
The PI3KdD946A protein is catalytically inactive mutant of PI3Kd with D946A mutation in the ATP binding site. This recombinant catal ytically inactive protein can be used as negative control in any kind of PI3Kd kinase activity studies. Reco 1 * 2,5 µG
Catalog # BIRBORB80846-2.5
Supplier:  Biorbyt
The PI3KdD946A protein is catalytically inactive mutant of PI3Kd with D946A mutation in the ATP binding site. This recombinant catal ytically inactive protein can be used as negative control in any kind of PI3Kd kinase activity studies. Reco 1 * 2,5 µG
Catalog # BIRBORB80846-2.5
Supplier:  Biorbyt