Insulin-Like Growth Factor mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids and having a molecular mass of 7600 Dalton. IGF-I is purified by proprietary chromatographic techniqu 1 * 50 µG
Catalog # BIRBORB80397-50
Supplier: Biorbyt
Frequently Bought Together