ContactUsLinkComponent
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 294 amino ac ids and having a molecular mass of 32.7 kDa. Recombinant Human Omentin contains His tag fused at N-Terminus. 1 * 2 µG
Catalog # BIRBORB80534-2
Supplier:  Biorbyt
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 294 amino ac ids and having a molecular mass of 32.7 kDa. Recombinant Human Omentin contains His tag fused at N-Terminus. 1 * 2 µG
Catalog # BIRBORB80534-2
Supplier:  Biorbyt