FGF 2 Human Recombinant (FGF-2) produced in Sf9 insect cells is a single, glycosylated, polypeptide chain containing 155 amino acids and having a molecular mass of 17353 Dalton.The FGF-basic is purified by proprietary chromatographic techni 1 * 2 µG
Catalog # BIRBORB80309-2
Supplier: Biorbyt
Frequently Bought Together