CD105 Human Recombinant extracellular domain produced in E.coli is a single, glycosylated, Polypeptide containing 151 amino acids (2 6-176) and having a molecular mass of 43 kDa.The CD-105 is purified by proprietary chromatographic technique 1 * 5 µG
BIRBORB80304-5
: Biorbyt
Frequently Bought Together