Oxytocin Human Synthetic is a single, non-glycosylated, polypeptide chain containing amino acids and having a molecular mass of 1007 .2144 Dalton.The OT is purified by proprietary chromatographic techniques. 1 * 25 mg
Catalog # BIRBORB80746-25
Supplier: Biorbyt
Frequently Bought Together