Lypressin is hormone used to prevent or control the frequent urination, increased thirst, and loss of water associated with diabetes insipidus (water diabetes). Its molecular weight is 1056.2 Dalton having an amino acid sequence of Cys-Tyr- 1 * 5 mg
Catalog # BIRBORB80740-5
Supplier: Biorbyt
Frequently Bought Together