ContactUsLinkComponent
Crosslab ptfe seal kit, for real seal wash systems, contains 5 ptfe o-ring seals 9mm x 1.5mm, 5 ptfe o-ring seals 32mm x 1.5mm, for ultimate 3 sd, rs, bm, and bx peristaltic pumps, tmo 1 * 1 items
Catalog # HEWL8002-0915
Supplier:  Agilent
Crosslab ptfe seal kit, for real seal wash systems, contains 5 ptfe o-ring seals 9mm x 1.5mm, 5 ptfe o-ring seals 32mm x 1.5mm, for ultimate 3 sd, rs, bm, and bx peristaltic pumps, tmo 1 * 1 items
Catalog # HEWL8002-0915
Supplier:  Agilent
Frequently Bought Together