ContactUsLinkComponent
Buffer loop 1ul wps-3sl/rs 1pk tmo crosslab buffer loop, 1ul, stainless steel, 1.6mm od x .5mm id x 52mm l, with fittings, for wps-3 sl analytical and wps-3rs, 1/pk, tmo 1 * 1 items
Catalog # HEWL8002-0860
Supplier:  Agilent
undefined
Buffer loop 1ul wps-3sl/rs 1pk tmo crosslab buffer loop, 1ul, stainless steel, 1.6mm od x .5mm id x 52mm l, with fittings, for wps-3 sl analytical and wps-3rs, 1/pk, tmo 1 * 1 items
Catalog # HEWL8002-0860
Supplier:  Agilent
Frequently Bought Together