ContactUsLinkComponent
Ndl seat capillary .5mmid 24ul 1pk tmo crosslab needle seat capillary,stainless steel, .8mm od x .5mm id x 12mm l, 24ul, with fittin gs, for ultimate 3 wps-3sl semiprep, 1/pk, tmo 1 * 1 items
Catalog # HEWL8002-0810
Supplier:  Agilent
Ndl seat capillary .5mmid 24ul 1pk tmo crosslab needle seat capillary,stainless steel, .8mm od x .5mm id x 12mm l, 24ul, with fittin gs, for ultimate 3 wps-3sl semiprep, 1/pk, tmo 1 * 1 items
Catalog # HEWL8002-0810
Supplier:  Agilent
Frequently Bought Together