ContactUsLinkComponent
Ndl seat capilry .12mmid 1.35ul 1pk tmo crosslab needle seat capillary, stainless steel, .8mm od x .12mm id  x 12mm l, 1.35ul, with fittings, for ultimate 3 wps-3sl, 1/pk, tmo 1 * 1 items
  HEWL8002-0808
 :  Agilent
undefined
Ndl seat capilry .12mmid 1.35ul 1pk tmo crosslab needle seat capillary, stainless steel, .8mm od x .12mm id  x 12mm l, 1.35ul, with fittings, for ultimate 3 wps-3sl, 1/pk, tmo 1 * 1 items
  HEWL8002-0808
 :  Agilent
Frequently Bought Together