ContactUsLinkComponent
Analog heavy_duty vortex mxier has control knob orbit 4,9mm. supplied w/ universal holder, cover, foam insert for (38) 1,5-2,0ml mic rotubes, cup head detachable 234cm powercord w/ ch style plug. 1 * 1 items
Catalog # 444-7135
Supplier:  VWR Collection
undefined
Analog heavy_duty vortex mxier has control knob orbit 4,9mm. supplied w/ universal holder, cover, foam insert for (38) 1,5-2,0ml mic rotubes, cup head detachable 234cm powercord w/ ch style plug. 1 * 1 items
Catalog # 444-7135
Supplier:  VWR Collection