Orders LinkContactUsLinkComponent
J.T.Baker®, Liquid Level Indicators
J.T.Baker®, Liquid Level Indicators
CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
Catalog # 9803-01
Supplier:   Avantor
J.T.Baker®, Liquid Level Indicators
Catalog # 9803-01
Supplier:  Avantor
J.T.Baker®, Liquid Level Indicators
CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits
Specifications
  • Description:
    CYCLE-TAINER™ LCD liquid level volume display, for 52 L model C system
  • Seq:
    02