ContactUsLinkComponent
Silica x 5µm particle size x 120Å pore size, 50mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
  SGEA2SIL-05L46P
Silica x 5µm particle size x 120Å pore size, 50mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
  SGEA2SIL-05L46P
Frequently Bought Together