ContactUsLinkComponent
Savon Soft care Sensisept 800ml 7516731 Mild liquid for washing and sanitizing Based on amphoteric and chlorhexidine digluconate Contains moisturizers and emollients without perfume or dye, dermatologically tested Disinfectant according to NF EN 1040 1 * 6 items
Catalog # DIVY7516731
Supplier:  Diversey
Savon Soft care Sensisept 800ml 7516731 Mild liquid for washing and sanitizing Based on amphoteric and chlorhexidine digluconate Contains moisturizers and emollients without perfume or dye, dermatologically tested Disinfectant according to NF EN 1040 1 * 6 items
Catalog # DIVY7516731
Supplier:  Diversey
Frequently Bought Together