Vacuum cleaner MRV-16(4W) #110800B Wet/Dry vacuum cleaner,intercepting tank stainless steel, weight:40 Kg power: 1050 watt Scope of delivery:Plastic equipment ( suction nozzle, round tool etc.)hose, filter bags. Tank including the casters are auto 1 * 1 Each
Catalog # TIGV110800B
Supplier: TIGER-VAC
Frequently Bought Together