Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
ESD CF 4 Sicherheitsschuhe, Blau EN ISO 20345 S1 Spitzenschutz Sicherheitsreflektoren aktiv-X Funktionsfutter atmungsaktive ESD Einlegesohle PPS-Dämpfungssystem MPU Sohlentechnologie 1 * 1 PAAR
ATLAESDCF4BLAU/41
:
ATLAS - the shoe company
ESD CF 4 Sicherheitsschuhe, Blau EN ISO 20345 S1 Spitzenschutz Sicherheitsreflektoren aktiv-X Funktionsfutter atmungsaktive ESD Einlegesohle PPS-Dämpfungssystem MPU Sohlentechnologie 1 * 1 PAAR
ATLAESDCF4BLAU/41
:
ATLAS - the shoe company
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
VWR® Facial Tissues
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection