DE 7516049 70% 5L 4cans ABBOTT 1lot Sterile surface disinfectant,70% Ethanol,30% Water for injection and 10ppm Bitrex,0,2µm filtered, gamma irradiated,double bagged, 5 L can., packaging unit 4x5 L 1 * 5 l
Catalog # DIVY7516049_U
Supplier: Diversey
Frequently Bought Together