B ClearKlensIPA sterilIPA70%300ml7514935 Sterile disinfectant, aerosol, 70% Isopropyl alcohol and 30% Water(purified water), 0,2µm filtered, gamma irradiated, double bagged, 300 ml aerosol bottle, packaging unit 12 x 300 ml 1 * 3,6 l
Catalog # 141-0888
Supplier: Diversey
Frequently Bought Together