ContactUsLinkComponent
Biotix Plate Seal Tape, 100/case. 2mm optically clear plate seal offers the clarity essential in real-time reactions an aggressive seal perfeect for reducinh evaporation expeirenced during thermal cycling. Functional temperature range -40C to 120C. 1 * 100 items
Catalog # 732-2490
Supplier:  BIOTIX
Biotix Plate Seal Tape, 100/case. 2mm optically clear plate seal offers the clarity essential in real-time reactions an aggressive seal perfeect for reducinh evaporation expeirenced during thermal cycling. Functional temperature range -40C to 120C. 1 * 100 items
Catalog # 732-2490
Supplier:  BIOTIX
Frequently Bought Together