Biotix Silver Seal Tape, 100/case. Aluminum sealing tape Ideal for PCR and storage. Chemically resistant but soft enough to conform to the plate and pierceable for robotic systems. Functional temperatrure range -70C-to 120 C. 1 * 100 items
732-2489
: BIOTIX
Frequently Bought Together