Digestion system, behrotest®
Hydrolysis unit, complete for one sample, HYDRO 1
Catalog # BEHRB00218430
Supplier: BEHR
Jump to:
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Specifications
- Description:Hydrolysis unit, complete for one sample
- Seq:01
- Type:HYDRO 1