Biotix 10 µL Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 20 Cards/Pack, 5 Pack/Case. Patented features: FlexF it™ for universal fit and superior seal, Chem-Resin™ for drip-free pipetting of polar solvents, and Blade™ for accuracy 1 * 1.920 items
Catalog # 613-2844
Supplier: BIOTIX
Frequently Bought Together