ContactUsLinkComponent
Biotix 10ul Pipette Tips. Racked, 10 racks of 96 Tips/Pack, 5 Packs/Case. Universal fit graduated, Patented features: FlexFit™ fo r universal fit and superior seal, Chem-Resin™ for drip-free pipetting of polar solvents, and Blade™ for accuracy 1 * 960 items
Catalog # 613-2831
Supplier:  BIOTIX
Biotix 10ul Pipette Tips. Racked, 10 racks of 96 Tips/Pack, 5 Packs/Case. Universal fit graduated, Patented features: FlexFit™ fo r universal fit and superior seal, Chem-Resin™ for drip-free pipetting of polar solvents, and Blade™ for accuracy 1 * 960 items
Catalog # 613-2831
Supplier:  BIOTIX
Frequently Bought Together