Speciation Standard: Arsenic(V) solution 1000 mg/L in Water 1 * 100 mL
Catalog # ACSDSP072.W.L1
Supplier: CPACHEM
Speciation Standard: Arsenic(V) solution 1000 mg/L in Water 1 * 100 mL
Catalog # ACSDSP072.W.L1
Supplier: CPACHEM
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Hydrochloric acid 2 mol/l (2 N) Reag. Ph. Eur. 1043503
Supplier: VWR Chemicals
VWR®, TOC standard solutions
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
Benzo(a)anthracene D12 certified 1 * 50 mg
Supplier: EHRENSTORFER
Camphene 1000 µg/mL in Isopropanol 1 * 1 mL
Supplier: EHRENSTORFER
VWR® PESTINORM®, 1,4-Dioxane, Stabilised, Secondary Reference Standard for GC
Supplier: VWR Chemicals
VWR®, Sulphur, Certified Reference Materials, in light mineral oil
Supplier: VWR Chemicals
MOXIDECTIN-D3 1 * 1 mg
Supplier: LGC Standards PROMOCHEM
Trifluralin D14 (di-n-propyl D14) 100 ng/µl 1 * 1,1 mL
Supplier: EHRENSTORFER
Eicosapentaenoic Acid, Reference Standards
Supplier: USP
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
Polyethylene Terephthalate, Reference Standards
Supplier: USP
Orion™ Conductivity Standards and Solutions, Thermo Scientific
Supplier: Thermo Orion
pH buffer solutions, colour coded
Supplier: Hach