Orders LinkContactUsLinkComponent
Viscosity flow cups
Viscosity flow cups
Viscosity flow cup, 6
Catalog # SIMEVF2197
Supplier:   Industrial Physics
Viscosity flow cups
Catalog # SIMEVF2197
Supplier:  Industrial Physics
Viscosity flow cups
Viscosity flow cups
Viscosity flow cup, 6
Price
191,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certifications: AFNOR
Documents
Specifications
  • Model:
    6
  • Description:
    Viscosity flow cup
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Temperature control jacket with tripod for ISO and AFNOR cups
    Temperature control jacket with tripod for ISO and AFNOR cups
    SIMEVF2068
    Log in for availability
    1 * 1 items - 627,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Tripod stand, 260 mm
    Tripod stand, 260 mm
    769-1150
    Log in for availability
    1 * 1 items - 217,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Digital stopwatch, 0,01 s
    Digital stopwatch, 0,01 s
    SIMEDI0076
    Log in for availability
    1 * 1 items - 29,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Ring stand fro flow cups
    Ring stand fro flow cups
    SIMEVF2062
    Log in for availability
    1 * 1 items - 83,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Viscosity convertor of flow time to viscosity
    Viscosity convertor of flow time to viscosity
    769-1151
    Log in for availability
    1 * 1 items - 87,50 €