ContactUsLinkComponent
pH combination electrode with Memosens plug head (ATEX II 1/G) Liquid electrolyte electrode with NTC 30K and KCl vessel 130 mm, Glas s shaft, ceramic junction, electrolyte L 310, Ag/AgCl-reference system, cylinder membrane, S glass, length 250 mm, 12 1 * 1 items
Catalog # SCOI285128350
Supplier:  SI Analytics
pH combination electrode with Memosens plug head (ATEX II 1/G) Liquid electrolyte electrode with NTC 30K and KCl vessel 130 mm, Glas s shaft, ceramic junction, electrolyte L 310, Ag/AgCl-reference system, cylinder membrane, S glass, length 250 mm, 12 1 * 1 items
Catalog # SCOI285128350
Supplier:  SI Analytics
Frequently Bought Together