ContactUsLinkComponent
pH combination electrode with screw plug head, Low maintenance with Pt 1000, glass shaft, 3 x ceramic diaphragm, gel electrolyte, Si lamid®-reference system, sphere membrane, A-glass, VP screw plug head, length 120 mm, 12 mm Ø, -5...+80 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285114790
Supplier:  SI Analytics
pH combination electrode with screw plug head, Low maintenance with Pt 1000, glass shaft, 3 x ceramic diaphragm, gel electrolyte, Si lamid®-reference system, sphere membrane, A-glass, VP screw plug head, length 120 mm, 12 mm Ø, -5...+80 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285114790
Supplier:  SI Analytics
Frequently Bought Together