ContactUsLinkComponent
Metal combination electrode with Memosens screw plug head (ATEX II 1/G) Low maintenance redox-combination electrode with NTC 30K, gl ass shaft, KPG annular gap junction, Referid-electrolyte, Silamid-ref., sensor platinum disk 6 mm Ø, length 120 mm, 1 1 * 1 items
Catalog # SCOI285111070
Supplier:  SI Analytics
Metal combination electrode with Memosens screw plug head (ATEX II 1/G) Low maintenance redox-combination electrode with NTC 30K, gl ass shaft, KPG annular gap junction, Referid-electrolyte, Silamid-ref., sensor platinum disk 6 mm Ø, length 120 mm, 1 1 * 1 items
Catalog # SCOI285111070
Supplier:  SI Analytics
Frequently Bought Together