ContactUsLinkComponent
Molsieve 13X, 80/100, preconditioned, 41.28 mm coil, 6.56 feet (2 m) x 1/16 inch, 2.1 mm, UltiMetal, packed GC column, for Large Val ve Oven 1 * 1 items
  HEWLG3591-81214
 :  Agilent
undefined
Molsieve 13X, 80/100, preconditioned, 41.28 mm coil, 6.56 feet (2 m) x 1/16 inch, 2.1 mm, UltiMetal, packed GC column, for Large Val ve Oven 1 * 1 items
  HEWLG3591-81214
 :  Agilent
Frequently Bought Together