ContactUsLinkComponent
Molsieve 5A, 60/80, preconditioned, 41.28 mm coil, 7 feet (2.13 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81209
Supplier:  Agilent
undefined
Molsieve 5A, 60/80, preconditioned, 41.28 mm coil, 7 feet (2.13 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81209
Supplier:  Agilent
Frequently Bought Together