Wipes polycontrol 900. high absorbancy and spill pich-up Wipes. cleanroom laundered and packed. sealed edges via laser cutting.ultr a low level of particles and extractables. unique two-ply construction for excellent absorbency 9x9in 1 * 1.000 items
Catalog # VILE142876
Supplier: FHP VILEDA SCS
Frequently Bought Together