ContactUsLinkComponent
Container to collect fluids. Capacity 1000 L. Raw material : high density polyethylene. Colour : yellow. To collect leak or overflow of dangerous fluids. 1 * 1 items
  LAMA3298
 :  LAMAPLAST
Container to collect fluids. Capacity 1000 L. Raw material : high density polyethylene. Colour : yellow. To collect leak or overflow of dangerous fluids. 1 * 1 items
  LAMA3298
 :  LAMAPLAST
Frequently Bought Together