ContactUsLinkComponent
1mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (ROTCW), 1mg/L (RTOCS02) C TOC as Sucrose and 1 mg/L (RTICN02) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120003
Supplier:  REAGECON
1mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (ROTCW), 1mg/L (RTOCS02) C TOC as Sucrose and 1 mg/L (RTICN02) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120003
Supplier:  REAGECON
Frequently Bought Together