Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Fine tip, 1 - 200 µl, Transparent, 2 racks, 96 each
Catalog # EDVO638
Supplier:   EDVOTEK
Pipette tips
Catalog # EDVO638
Supplier:  EDVOTEK
Pipette tips
Pipette tips
Fine tip, 1 - 200 µl, Transparent, 2 racks, 96 each
Price
38,50 €
Log in for availability
1 * 204 items
Product Details & Documents
Specifications
  • Packed:
    2 racks, 96 each
  • Volume:
    1 - 200 µl
  • Tip style:
    Fine tip
  • Tip colour:
    Transparent