Spectrum analyzer, frequency range 9KHz 3GHz, high frequency stability 25ppb (0.025ppm), phase noise: -88dbc/hz@1ghz, 10khz offset , built-in measurement functions: channel power, n-db bandwidth, OCBW, ACPR, SEM, TOI, CNR, CTB, 1 * 1 items
Catalog # DIDAPMD500010_UK
Supplier: DIDALAB
Frequently Bought Together