ContactUsLinkComponent
RLC meter, wide test frequency: 20Hz 5MHz, 0.1% basic accuracy and 6 digits measurement, large LCD display with intuitive user int erface, full measuring functions with dut V/I. 1 * 1 items
  DIDAPMD400120
 :  DIDALAB
RLC meter, wide test frequency: 20Hz 5MHz, 0.1% basic accuracy and 6 digits measurement, large LCD display with intuitive user int erface, full measuring functions with dut V/I. 1 * 1 items
  DIDAPMD400120
 :  DIDALAB
Frequently Bought Together