ContactUsLinkComponent
RLC meter, wide test frequency 20Hz 1MHz, R 0.1M 100M, basic accuracy and 6 digits measurement and large lcd display with intu itive user interface and full measuring functions with dut V/I. 1 * 1 items
Catalog # DIDAPMD400110
Supplier:  DIDALAB
RLC meter, wide test frequency 20Hz 1MHz, R 0.1M 100M, basic accuracy and 6 digits measurement and large lcd display with intu itive user interface and full measuring functions with dut V/I. 1 * 1 items
Catalog # DIDAPMD400110
Supplier:  DIDALAB
Frequently Bought Together