Orders LinkContactUsLinkComponent
Optical components for optical bench
Optical components for optical bench
Set of filters for Michelson interferometry for use with CCD Caliens camera
Catalog # DIDAPOD010025
Supplier:   DIDALAB
Optical components for optical bench
Catalog # DIDAPOD010025
Supplier:  DIDALAB
Optical components for optical bench
Optical components for optical bench
Set of filters for Michelson interferometry for use with CCD Caliens camera
Price
162,75 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Set of filters for Michelson interferometry for use with CCD Caliens camera
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Permanent frame, Ø 40 mm
    Permanent frame, Ø 40 mm
    DIDAPOD010095
    Log in for availability
    1 * 1 items - 42,87 €
Product Information
  • Catalog #
    Availability
    Price
  • Simple lens holder, Ø 40 mm
    Simple lens holder, Ø 40 mm
    DIDAPOD010110
    Log in for availability
    1 * 1 items - 21,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lens holder for fragile components, Ø 80 mm
    Lens holder for fragile components, Ø 80 mm
    DIDAPOD010080
    Log in for availability
    1 * 1 items - 75,25 €
Product Information
  • Catalog #
    Availability
    Price
  • Lens holder, Ø 40/42 mm
    Lens holder, Ø 40/42 mm
    DIDAPOD010090
    Log in for availability
    1 * 1 items - 39,38 €
Product Information
  • Catalog #
    Availability
    Price
  • Lens holder, Ø 80 mm
    Lens holder, Ø 80 mm
    DIDAPOD060130
    Log in for availability
    1 * 1 items - 60,38 €
Product Information
  • Catalog #
    Availability
    Price
  • Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    DIDAPOD002192
    Log in for availability
    1 * 1 items - 126,00 €