Oscilloscope digital, bandwidth 50MHz, 2channels, up to 19 auto measurements, FFT-RMS, zoom FFT, math functions +, -, x, FFT, 5.7inc hes colour TFT LCD screen, USB host and device ports, go/no-go function, data logger. 1 * 1 items
DIDAPMD100090
: DIDALAB
Frequently Bought Together