ContactUsLinkComponent
Oscilloscope digital, UK, bandwidth 70MHz, 2channels, up to 27 auto measurements, FFT-RMS, zoom FFT, math functions +, -, x, FFT, 5. 7inches colour TFT LCD screen, USB host and device ports, go/no-go function, data logger. 1 * 1 items
Catalog # DIDAPMD100060_UK
Supplier:  DIDALAB
Oscilloscope digital, UK, bandwidth 70MHz, 2channels, up to 27 auto measurements, FFT-RMS, zoom FFT, math functions +, -, x, FFT, 5. 7inches colour TFT LCD screen, USB host and device ports, go/no-go function, data logger. 1 * 1 items
Catalog # DIDAPMD100060_UK
Supplier:  DIDALAB
Frequently Bought Together