ContactUsLinkComponent
Generator, arbitrary functions, UK, 0.1Hz 50MHz with 0.1Hz resolution, AM/FM/FSK modulation-sweep and frequency counter, sine, squ are, ramp, noise , arbitrary wavefrom signals, 1%99% adjustable duty cycle, 3.5 inches LCD screen, U 1 * 1 items
Catalog # DIDAPMD200040-UK
Supplier:  DIDALAB
Generator, arbitrary functions, UK, 0.1Hz 50MHz with 0.1Hz resolution, AM/FM/FSK modulation-sweep and frequency counter, sine, squ are, ramp, noise , arbitrary wavefrom signals, 1%99% adjustable duty cycle, 3.5 inches LCD screen, U 1 * 1 items
Catalog # DIDAPMD200040-UK
Supplier:  DIDALAB
Frequently Bought Together