ContactUsLinkComponent
Michelson interferometer, motorized, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of t he beamsplitters and mirrors /20, we put the orientation and lead systems of the compensator b . 1 * 1 items
Catalog # DIDAPOD013493_UK
Supplier:  DIDALAB
Michelson interferometer, motorized, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of t he beamsplitters and mirrors /20, we put the orientation and lead systems of the compensator b . 1 * 1 items
Catalog # DIDAPOD013493_UK
Supplier:  DIDALAB
Frequently Bought Together