Gratings transmission, 10 lines/mm, these accuracy gratings have a great uniformity of lines on the whole working area, the working area (36x24mm) is protected by a glass plate, compatible with our slide-holders. 1 * 1 items
DIDAPOD062500
: DIDALAB
Frequently Bought Together