ContactUsLinkComponent
Telescope, autocollimating, high optical quality, achromatic antiglare lens, high quality team splitter, reticle with adjustable cro ss, with a knurled ring, very large focal length of 162mm, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD068101
Supplier:  DIDALAB
Telescope, autocollimating, high optical quality, achromatic antiglare lens, high quality team splitter, reticle with adjustable cro ss, with a knurled ring, very large focal length of 162mm, mounted on diameter 10mm rod. 1 * 1 items
Catalog # DIDAPOD068101
Supplier:  DIDALAB
Frequently Bought Together