Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Dipslide Dermatest, 10
Catalog # LIOF50021
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF50021
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Dipslide Dermatest, 10
CAS Number:  
Price
27,30 €
Log in for availability
1 * 10 items
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Specifications
Specifications
  • Pk:
    10
  • Description:
    Dipslide Dermatest