Luthetium Lu - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM331.2NP.L1
Supplier: CPACHEM
Luthetium Lu - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM331.2NP.L1
Supplier: CPACHEM
Frequently Bought Together
ARIPIPRAZOLE-D8 1 * 1 mg
Supplier: LGC Standards PROMOCHEM
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion
Platinum standard solution, 1000 mg/L Pt in 20% hydrochloric acid (from Pt), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
1-Bromo-2-chloroethane
Supplier: EHRENSTORFER
Aloin, Reference Standard
Supplier: USP
CHLOROBUTANOL HEMIHYDRATE
Supplier: EDQM
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Paracetamol
Supplier: EDQM
BUDESONIDE 9(11)-ENE 1 * 25 mg
Supplier: Merck
Parsol 340 certified 1 * 0,25 g
Supplier: EHRENSTORFER
SULFUR 200 µG/G IN ISOOCTANE 1 * 100 mL
Supplier: AccuStandard