Orders LinkContactUsLinkComponent
Multi-element standards for ICP
Multi-element standards for 33 elements, 5% HNO₃, 500 mL
Catalog # ACSDM8A96.K1.5N.L5
Supplier:   CPACHEM
Multi-element standards for ICP
Catalog # ACSDM8A96.K1.5N.L5
Supplier:  CPACHEM
CAS Number:  
Multi-element standards for ICP
Multi-element standards for 33 elements, 5% HNO₃, 500 mL
CAS Number:  
Price
942,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
ICP standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn.
Specifications
Specifications
  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    Multi-element standards for 33 elements
Specifications