ContactUsLinkComponent
CALCOCO1 (Human) Recombinant protein (P01). Human CALCOCO1 full-length ORF ( NP_065949.1, 1 A.A. - 691 A.A.) Recombinant protein wit h GST-TAG AT N-terminal. 1 * 25 G
Catalog # ABNOH00057658-P012
Supplier:  Abnova
CALCOCO1 (Human) Recombinant protein (P01). Human CALCOCO1 full-length ORF ( NP_065949.1, 1 A.A. - 691 A.A.) Recombinant protein wit h GST-TAG AT N-terminal. 1 * 25 G
Catalog # ABNOH00057658-P012
Supplier:  Abnova
Frequently Bought Together