Calibration gas 25 h2s,100 co,0.85%c3h8,18% o2,n2 4g18x85w25h100-34 1 * 1 items
BWTE4G18X85W25H100
Calibration gas 25 h2s,100 co,0.85%c3h8,18% o2,n2 4g18x85w25h100-34 1 * 1 items
BWTE4G18X85W25H100
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection